SP Wilmad-LabGlass Spherical Half-Jacketed Reaction Vessels, Conical Flange, SP Industries
ML-1284-700 ML-1284-704 LG-8022-098 LG-8022-102 ML-1284-702 LG-8022-104
14205-230EA 1258.73 USD
14205-230 14206-636 14209-086 14207-358 14206-892 14204-474
SP Wilmad-LabGlass Spherical Half-Jacketed Reaction Vessels, Conical Flange, SP Industries
These borosilicate glass reaction vessels are half-jacketed with 1/2” hose connections for the circulating heating or cooling medium.
Top is finely ground to provide for a leak-proof fit with lids LG-8030 to LG-8050, ML-1285, and ML-1286.
Ordering information: ML-1288-700 clamp is used with the 200 mL to 500 mL flask; LG-8070-100 clamp is used with the 2000 mL to 1200 mL flasks. (Not included.)