L-Serine, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT910-250UCI MT910-1MCI MT910-5MCI
101973-922EA 2576.15 USD
101973-922 101973-926 101973-924
L-Serine, [3H] ≥97% (by HPLC), MORpure™
L-Serine, [3H]
CAS Number: 83245-15-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR