2'-Deoxyguanosine-5'-triphosphate, tetraammonium salt, [8-3H] ≥96% (by HPLC)

MT884-5MCI MT884-250UCI MT884-1MCI
101973-822EA 5378.26 USD
101973-822 101973-820 101973-818
2'-Deoxyguanosine-5'-triphosphate, tetraammonium salt, [8-3H] ≥96% (by HPLC)
2'-Deoxyguanosine-5'-triphosphate, tetraammonium salt, [8-3H]
CAS Number: 340130-25-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR