L-(+)-Arabinose, [1-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT851-250UCI MT851-1MCI MT851-5MCI
101973-788EA 3347.69 USD
101973-788 101973-786 101973-790
L-(+)-Arabinose, [1-3H(N)] ≥97% (by HPLC), MORpure™
L-(+)-Arabinose, [1-3H(N)]
CAS Number: 90865-52-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR