Thymidine (2'-deoxythymidine)
Supplier: MORAVEK BIOCHEMICALS MS
|
|
Synonyms:
dT
, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione
, Thymine deoxyriboside
MT846-1MCI
MT846-250UCI
MT846-5MCI
101973-756EA
3746.54
USD
101973-756
101973-754
101973-752
Thymidine (2'-deoxythymidine)
Thymidine (2'-Deoxythymidine)
Formula:
C₁₀H₁₄N₂O₅ MW: 242.23 g/mol |
MDL Number:
MFCD00006537 CAS Number: 50-89-5 UN: 2910 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...