2',3'-Dideoxyguanosine, [2',3'-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT829-250UCI
101973-712EA 2576.15 USD
101973-712
2',3'-Dideoxyguanosine, [2',3'-3H(N)] ≥97% (by HPLC), MORpure™
2',3'-Dideoxyguanosine, [2',3'-3H(N)]
CAS Number: 339532-37-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR