N-Methyl-N-nitrosourea, [3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT813-1MCI MT813-5MCI
101973-678EA 7081.15 USD
101973-678 101973-676
N-Methyl-N-nitrosourea, [3H] ≥95% (by HPLC)
N-Methyl-N-nitrosourea, [3H]
CAS Number: 2086337-10-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR