5-Fluorocytosine, [6-3H] ≥97% (by HPLC), MORpure™

MT811-250UCI
101973-664EA 2171.14 USD
101973-664
5-Fluorocytosine, [6-3H] ≥97% (by HPLC), MORpure™
5-Fluorocytosine, [6-3H]
CAS Number: 132567-20-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR