Folic acid, diammonium salt, [3',5',7,9-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT783K-500UCI MT783K-250UCI MT783K-50UCI MT783K-1MCI
101973-602EA 3158.08 USD
101973-602 101973-600 101973-606 101973-604
Folic acid, diammonium salt, [3',5',7,9-3H] ≥97% (by HPLC), MORpure™
Folic acid, diammonium salt, [3',5',7,9-3H]
CAS Number: 339529-03-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR