Cytidine-5'-monophosphate, diammonium salt, [5-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT762-250UCI
101973-560EA 2576.15 USD
101973-560
Cytidine-5'-monophosphate, diammonium salt, [5-3H] ≥97% (by HPLC), MORpure™
Cytidine-5'-monophosphate, diammonium salt, [5-3H]
CAS Number: 339527-42-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR