Inosine, [2,8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT678-250UCI MT678-1MCI
101973-388EA 2576.15 USD
101973-388 101973-386
Inosine, [2,8-3H] ≥97% (by HPLC), MORpure™
Inosine, [2,8-3H]
CAS Number: 82252-19-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR