Vitamin K3, [3H]

Supplier: MORAVEK BIOCHEMICALS MS

MT664-250UCI
101973-326EA 2948.85 USD
101973-326
Vitamin K3, [3H]
Vitamin K3, [3H]
CAS Number: 84979-90-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR