Vinblastine sulfate, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT657S-50UCI MT657S-250UCI
101973-320EA 4308.85 USD
101973-320 101973-318
Vinblastine sulfate, [3H] ≥97% (by HPLC), MORpure™
Vinblastine sulfate, [3H]
CAS Number: 57532-77-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR