ACVMP, [8-3H] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT652-250UCI
101973-300EA 2576.15 USD
101973-300
ACVMP, [8-3H] ≥95% (by HPLC)
ACVMP, [8-3H]
CAS Number: 339340-51-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR