5-Iodouracil, [6-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT627-250UCI
101973-240EA 2576.15 USD
101973-240
5-Iodouracil, [6-3H] ≥97% (by HPLC), MORpure™
5-Iodouracil, [6-3H]
CAS Number: 36120-39-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR