Riboflavin, [3H] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT605-1MCI MT605-250UCI
101973-180EA 3543.85 USD
101973-180 101973-178
Riboflavin, [3H] ≥96% (by HPLC)
Riboflavin, [3H]
CAS Number: 42880-33-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR