Uridine, [5-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT602E-1MCI MT602E-5MCI MT602E-250UCI
101973-118EA 523.08 USD
101973-118 101973-122 101973-120
Uridine, [5-3H] ≥97% (by HPLC), MORpure™
Uridine, [5-3H]
CAS Number: 3705-45-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR