Uridine, [5-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT602-5MCI MT602-250UCI MT602-1MCI
101973-116EA 2903.08 USD
101973-116 101973-114 101973-112
Uridine, [5-3H] ≥97% (by HPLC), MORpure™
Uridine, [5-3H]
CAS Number: 3705-45-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR