Valproic acid, sodium salt, [3H] ≥97% (by HPLC), MORpure™

MT556-250UCI MT556-1MCI
101973-084EA 1146.18 USD
101973-084 101973-082
Valproic acid, sodium salt, [3H] ≥97% (by HPLC), MORpure™
Valproic acid, sodium salt, [3H]
CAS Number: 2086336-95-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR