Xanthine, [8-3H] ≥97% (by HPLC), MORpure™

MT537-250UCI MT537-1MCI
101973-018EA 2942.61 USD
101973-018 101973-016
Xanthine, [8-3H] ≥97% (by HPLC), MORpure™
Xanthine, [8-3H]
CAS Number: 54565-62-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR