L-Adenosine, [2,8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT530-1MCI
101973-010EA 1928.85 USD
101973-010
L-Adenosine, [2,8-3H] ≥97% (by HPLC), MORpure™
L-Adenosine, [2,8-3H]
CAS Number: 339158-15-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR