Estradiol, [2,4-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT524-250UCI
101973-004EA 2576.15 USD
101973-004
Estradiol, [2,4-3H] ≥97% (by HPLC), MORpure™
Estradiol, [2,4-3H]
CAS Number: 98722-94-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR