Ochratoxin A, [3H] ≥96% (by HPLC)

MT518-50UCI MT518-250UCI MT518-1MCI
101972-990EA 3300.79 USD
101972-990 101972-986 101972-988
Ochratoxin A, [3H] ≥96% (by HPLC)
Ochratoxin A, [3H]
CAS Number: 189152-21-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR