Aminopterin, [3',5',7,9-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT506-250UCI
101972-938EA 2576.15 USD
101972-938
Aminopterin, [3',5',7,9-3H(N)] ≥97% (by HPLC), MORpure™
Aminopterin, [3',5',7,9-3H(N)]
CAS Number: 339155-95-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR