Doxorubicin, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1963-50UCI
101972-884EA 1516.92 USD
101972-884
Doxorubicin, [3H] ≥97% (by HPLC), MORpure™
Doxorubicin, [3H]
CAS Number: 946832-45-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR