Cyclosporin A, [3H] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1961-250UCI
101972-876EA 2576.15 USD
101972-876
Cyclosporin A, [3H] ≥96% (by HPLC)
Cyclosporin A, [3H]
CAS Number: 225937-78-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR