8-Aminoadenosine, [2-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1926-250UCI
101972-808EA 2576.15 USD
101972-808
8-Aminoadenosine, [2-3H] ≥97% (by HPLC), MORpure™
8-Aminoadenosine, [2-3H]
CAS Number: 2086327-59-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR