2-BFI, [4,6-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1911-250UCI
101972-772EA 2870.38 USD
101972-772
2-BFI, [4,6-3H] ≥97% (by HPLC), MORpure™
2-BFI, [4,6-3H]
CAS Number: 2087960-09-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR