N-Palmitoylethanolamine, [ethanolamine-1,2-3H] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1866-250UCI
101972-662EA 2576.15 USD
101972-662
N-Palmitoylethanolamine, [ethanolamine-1,2-3H] ≥96% (by HPLC)
N-Palmitoylethanolamine, [ethanolamine-1,2-3H]
CAS Number: 2086337-01-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR