Abacavir

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-methanol((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol

MT1736-250UCI
101972-522EA 2576.15 USD
101972-522
Abacavir
Abacavir
Formula: C₁₄H₁₈N₆O
MW: 286.34 g/mol
MDL Number: MFCD00903850
CAS Number: 136470-78-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR