(S)-Warfarin, [phenyl-4-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1628-250UCI
101972-376EA 4106.15 USD
101972-376
(S)-Warfarin, [phenyl-4-3H] ≥97% (by HPLC), MORpure™
(S)-Warfarin, [phenyl-4-3H]
CAS Number: 339155-40-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR