NNK, [pyridyl-5-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1621-10UCI MT1621-50UCI MT1621-250UCI
101972-350EA 1170.38 USD
101972-350 101972-352 101972-354
NNK, [pyridyl-5-3H] ≥97% (by HPLC), MORpure™
NNK, [pyridyl-5-3H]
CAS Number: 117637-98-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR