D-Glutamic acid, [3H] ≥97% (by HPLC), MORpure™

MT1577-1MCI MT1577-250UCI
101972-286EA 1206.8 USD
101972-286 101972-288
D-Glutamic acid, [3H] ≥97% (by HPLC), MORpure™
D-Glutamic acid, [3H]
CAS Number: 2087435-30-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR