Diethylstilbestrol, [3H] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MT1570-250UCI
101972-270EA 3393.46 USD
101972-270
Diethylstilbestrol, [3H] ≥96% (by HPLC)
Diethylstilbestrol, [3H]
CAS Number: 316122-73-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR