Quinolinic acid, [3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1566-250UCI
101972-252EA 2576.15 USD
101972-252
Quinolinic acid, [3H] ≥97% (by HPLC), MORpure™
Quinolinic acid, [3H]
CAS Number: 339155-13-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR