5,7-Dihydroxy-4'-methoxyisoflavone, [6,8,3',5'-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1563-250UCI
101972-244EA 2576.15 USD
101972-244
5,7-Dihydroxy-4'-methoxyisoflavone, [6,8,3',5'-3H(N)] ≥97% (by HPLC), MORpure™
5,7-Dihydroxy-4'-methoxyisoflavone, [6,8,3',5'-3H(N)]
CAS Number: 316351-54-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR