2-Nitro-1H-imidazole, [2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC498-10UCI MC498-50UCI
101972-012EA 1621.54 USD
101972-012 101972-014
2-Nitro-1H-imidazole, [2-14C] ≥97% (by HPLC)
2-Nitro-1H-imidazole, [2-14C]
CAS Number: 99481-47-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR