Cytidine-5'-diphosphate, triammonium salt, [2-14C] ≥95% (by HPLC)

MC480-10UCI MC480-50UCI
101972-000EA 1366.6 USD
101972-000 101972-002
Cytidine-5'-diphosphate, triammonium salt, [2-14C] ≥95% (by HPLC)
Cytidine-5'-diphosphate, triammonium salt, [2-14C]
CAS Number: 337511-52-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR