Anisole, [ring-14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC477-50UCI
101971-988EA 5191.54 USD
101971-988
Anisole, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Anisole, [ring-14C(U)]
CAS Number: 53379-84-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR