Glycylsarcosine, [glycyl-1,2-14C] ≥97% (by HPLC)

MC447-10UCI MC447-25UCI MC447-50UCI
101971-916EA 1250.88 USD
101971-916 101971-914 101971-918
Glycylsarcosine, [glycyl-1,2-14C] ≥97% (by HPLC)
Glycylsarcosine, [glycyl-1,2-14C]
CAS Number: 2088008-46-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR