D,L-4,5-Dihydroorotic acid, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC420-10UCI
101971-888EA 1621.54 USD
101971-888
D,L-4,5-Dihydroorotic acid, [2-14C] ≥98% (by HPLC), MORpure™
D,L-4,5-Dihydroorotic acid, [2-14C]
CAS Number: 337332-11-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR