Butyryl coenzyme A [butyryl-1-14C] ≥96% (by HPLC), CAT Assay Grade

MC330-10UCI
101971-792EA 1096.59 USD
101971-792
Butyryl coenzyme A [butyryl-1-14C] ≥96% (by HPLC), CAT Assay Grade
Butyryl coenzyme A [butyryl-1-14C]
CAS Number: 289906-82-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR