D-threo-Chloramphenicol, [dichloroacetyl-1,2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC302-50UCI
101971-734EA 5191.54 USD
101971-734
D-threo-Chloramphenicol, [dichloroacetyl-1,2-14C] ≥98% (by HPLC), MORpure™
D-threo-Chloramphenicol, [dichloroacetyl-1,2-14C]
CAS Number: 289677-22-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR