Acetyl coenzyme A, [acetyl-1-14C] ≥98% (by HPLC), MORpure™, CAT Assay Grade

MC269L-10UCI MC269L-25UCI MC269L-50UCI
101971-670EA 837.6 USD
101971-670 101971-674 101971-672
Acetyl coenzyme A, [acetyl-1-14C] ≥98% (by HPLC), MORpure™, CAT Assay Grade
Acetyl coenzyme A, [acetyl-1-14C]
CAS Number: 4332-37-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR