Acetyl coenzyme A, [acetyl-1-14C] ≥98% (by HPLC), MORpure™

MC269-25UCI MC269-10UCI MC269-250UCI MC269-50UCI
101971-662EA 837.6 USD
101971-662 101971-666 101971-664 101971-668
Acetyl coenzyme A, [acetyl-1-14C] ≥98% (by HPLC), MORpure™
Acetyl coenzyme A, [acetyl-1-14C]
CAS Number: 4332-37-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR