Succinic acid, [2,3-14C] ≥98% (by HPLC), MORpure™

MC268-250UCI MC268-50UCI MC268-1MCI
101971-660EA 3521.21 USD
101971-660 101971-656 101971-658
Succinic acid, [2,3-14C] ≥98% (by HPLC), MORpure™
Succinic acid, [2,3-14C]
CAS Number: 5163-60-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR