Saccharin, [ring-14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2419-50UCI MC2419-10UCI
101971-586EA 5191.54 USD
101971-586 101971-584
Saccharin, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Saccharin, [ring-14C(U)]
CAS Number: 2087971-15-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR