Phenylacetic acid, [ring-14C(U)] ≥98% (by HPLC), MORpure™

MC2346-50UCI
101971-458EA 4375.34 USD
101971-458
Phenylacetic acid, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Phenylacetic acid, [ring-14C(U)]
CAS Number: 2088208-93-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR