FD&C blue #1, [aniline-14C(U)] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2342-10UCI
101971-454EA 1621.54 USD
101971-454
FD&C blue #1, [aniline-14C(U)] ≥97% (by HPLC)
FD&C blue #1, [aniline-14C(U)]
CAS Number: 2088057-69-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR