Formic acid, free acid, [14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2308-5MCI
101971-384EA 5204.62 USD
101971-384
Formic acid, free acid, [14C] ≥98% (by HPLC), MORpure™
Formic acid, free acid, [14C]
CAS Number: 1633-55-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR